Learn More
Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
128.00 € - 1823.00 €
Chemical Identifiers
CAS | 1313-82-2 |
---|---|
Molecular Formula | Na2S |
Molecular Weight (g/mol) | 78.04 |
MDL Number | MFCD00003498 |
InChI Key | GRVFOGOEDUUMBP-UHFFFAOYSA-N |
Synonym | sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s |
PubChem CID | 237873 |
SMILES | [Na+].[Na+].[S--] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10750971
|
Thermo Scientific Acros
310180100 |
10 g | Glass bottle |
128.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10618264
|
Thermo Scientific Acros
310180500 |
50 g | Glass bottle |
474.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13408518
|
Thermo Scientific Acros
310182500 |
250 g | Glass bottle |
1823.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-82-2 | |
78.04 | |
GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
237873 |
Na2S | |
MFCD00003498 | |
sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s | |
[Na+].[Na+].[S--] |
Specifications
1313-82-2 | |
Lump/Powder | |
Na2S | |
MFCD00003498 | |
sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s | |
GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
disodium;sulfanide | |
237873 | |
≥90% | |
Sodium sulfide, 90%, balance water |
Beige-Yellow | |
90% min. (Iodimetry) | |
Na2S | |
15,881 | |
Solubility in water: 186g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether | |
[Na+].[Na+].[S--] | |
78.04 | |
78.04 | |
Pure |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
Corrosive
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-211-5
missing translation for 'rtecsNumber' : WE1905000
missing translation for 'tsca' : TSCA
- Sodium sulfide
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- EUH031-Contact with acids liberates toxic gas.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only