Learn More
Sodium sulfide nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
35.80 € - 229.00 €
Chemical Identifiers
CAS | 1313-84-4 |
---|---|
Molecular Formula | H18Na2O9S |
Molecular Weight (g/mol) | 240.17 |
MDL Number | MFCD00149184 |
InChI Key | ZGHLCBJZQLNUAZ-UHFFFAOYSA-N |
IUPAC Name | disodium nonahydrate sulfanediide |
SMILES | O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10656811
|
Thermo Scientific Acros
424420250 |
25 g | Glass bottle | N/A | N/A | N/A | |||||
10587952
|
Thermo Scientific Acros
424425000 |
500 g | Glass Bottle | N/A | N/A | N/A | |||||
10015643
|
Thermo Scientific Acros
424420025 |
2.5 kg | Plastic bottle | N/A | N/A | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-84-4 | |
240.17 | |
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
H18Na2O9S | |
MFCD00149184 | |
disodium nonahydrate sulfanediide |
Specifications
50.0°C | |
Colorless or Yellow | |
98% min. (Iodimetry) | |
Na2S·9H2O | |
01,1104; 11,492 | |
Solubility in water: 180g/L (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether | |
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] | |
240.17 | |
≥98% | |
Sodium sulfide nonahydrate |
1313-84-4 | |
Adhering Crystals Chunks | |
H18Na2O9S | |
MFCD00149184 | |
15, 8810 | |
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
disodium nonahydrate sulfanediide | |
240.18 | |
ACS Reagent |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
Corrosiv
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : WE1925000
- Sodium sulfide nonahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H290-May be corrosive to metals.
- EUH031-Contact with acids liberates toxic gas.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.