missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetraborate decahydrate, Honeywell Fluka™
ACS Reagent,≥99.5%
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15688150
|
Honeywell-Fluka
S9640-25G |
25 g |
N/A
|
N/A | |||||
15608160
|
Honeywell-Fluka
S9640-500G |
500 g |
N/A
|
N/A | |||||
15678150
|
Honeywell-Fluka
S9640-2.5KG |
2.5 kg |
N/A
|
N/A | |||||
15618160
|
Honeywell-Fluka
S9640-6X500G |
6 x 500 g |
N/A
|
N/A | |||||
15698150
|
Honeywell-Fluka
S9640-4X2.5KG |
4 x 2.5 kg |
N/A
|
N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1303-96-4 | |
MFCD00149193 |
Specifications
1303-96-4 | |
MFCD00149193 | |
381.37g/mol | |
1.73g/mL at 25°C |
Na2B4O7 · 10H2O | |
NONH for all modes of transport | |
≥99.5% | |
Sodium tetraborate decahydrate |
Safety and Handling
P201-P280-P308 + P313
missing translation for 'rtecsNumber' : VZ2275000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium tetraborate decahydrate
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.