missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™
H10Na2O8S2, CAS Number-10102-17-7, ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m, 500g, CHEBI:32150, White
Chemical Identifiers
CAS | 10102-17-7 |
---|---|
Molecular Formula | H10Na2O8S2 |
Molecular Weight (g/mol) | 248.172 |
MDL Number | 149186 |
InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
PubChem CID | 61475 |
ChEBI | CHEBI:32150 |
IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
SMILES | O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10294250
|
Fisher Chemical
S/7240/53 |
500 g | Plastic powder jar |
N/A
|
N/A | ||||
10090350
|
Fisher Chemical
S/7240/60 |
1 kg | Plastic powder jar |
N/A
|
N/A | ||||
10192250
|
Fisher Chemical
S/7240/63 |
3 kg | Plastic powder jar |
N/A
|
N/A | ||||
10723811
|
Fisher Chemical
S/7240/65 |
5 kg | Plastic keg |
N/A
|
N/A | ||||
Description
H10Na2O8S2, CAS Number-10102-17-7, ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m, 500g, CHEBI:32150, White
Chemical Identifiers
10102-17-7 | |
248.172 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
61475 | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
H10Na2O8S2 | |
149186 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
CHEBI:32150 | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Specifications
48.5°C | |
White | |
Solid | |
149186 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
61475 | |
248.18g/mol |
10102-17-7 | |
6.0 to 8.4 | |
H10Na2O8S2 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
248.172 | |
CHEBI:32150 |