missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
69.55 € - 117.00 €
Chemical Identifiers
CAS | 7772-98-7 |
---|---|
Molecular Formula | Na2O3S2 |
Molecular Weight (g/mol) | 158.10 |
MDL Number | MFCD00003499 |
InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
PubChem CID | 61475 |
ChEBI | CHEBI:32150 |
IUPAC Name | disodium sulfanidesulfonate |
SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10243960
|
Fisher Chemical
J/7950/17 |
2.5 L | HDPE plastic bottle |
69.55 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10753694
|
Fisher Chemical
J/7950/21 |
5 L | HDPE plastic bottle |
73.60 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10121910
|
Fisher Chemical
J/7950/24 |
10 L | Bag in box |
117.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis
Chemical Identifiers
7772-98-7 | |
158.10 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
61475 | |
disodium sulfanidesulfonate |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
CHEBI:32150 | |
[Na+].[Na+].[O-]S([S-])(=O)=O |
Specifications
7772-98-7 | |
Na2O3S2 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
158.10 | |
CHEBI:32150 |
Liquid | |
MFCD00003499 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium sulfanidesulfonate | |
61475 |