Learn More
Sodium triacetoxyborohydride, 97%, Thermo Scientific Chemicals
67.50 € - 557.00 €
Chemical Identifiers
CAS | 56553-60-7 |
---|---|
Molecular Formula | C6H10BNaO6 |
Molecular Weight (g/mol) | 211.94 |
MDL Number | MFCD00012211 |
InChI Key | HHYFEYBWNZJVFQ-UHFFFAOYSA-N |
Synonym | c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate |
IUPAC Name | sodium bis(acetyloxy)boranuidyl acetate |
SMILES | [Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10184970
|
Thermo Scientific Acros
291820250 |
25 g | Glass bottle |
67.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10677431
|
Thermo Scientific Acros
291821000 |
100 g | Glass Bottle |
170.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10513094
|
Thermo Scientific Acros
291825000 |
500 g | Glass bottle |
557.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
56553-60-7 | |
211.94 | |
HHYFEYBWNZJVFQ-UHFFFAOYSA-N | |
sodium bis(acetyloxy)boranuidyl acetate |
C6H10BNaO6 | |
MFCD00012211 | |
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate | |
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O |
Specifications
56553-60-7 | |
White | |
96% min | |
NaB(O2CCH3)3H | |
12,453; 13,283; 06,553; 16,309 | |
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate | |
HHYFEYBWNZJVFQ-UHFFFAOYSA-N | |
sodium bis(acetyloxy)boranuidyl acetate | |
211.94 | |
Powder |
114°C to 118°C | |
Authentic | |
C6H10BNaO6 | |
MFCD00012211 | |
15, 8822 | |
Solubility in water: reacts. Other solubilities: soluble in thf,acetonitrile,1,2-dichloroethane | |
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O | |
211.94 | |
97% | |
Sodium triacetoxyborohydride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
- Sodium triacetoxyborohydride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
- P223-Do not allow contact with water.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402-Store in a dry place.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.