missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Black
Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.
28.75 € - 66.05 €
Specifications
Color | Black |
---|---|
Material | Microcellular Vinyl |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
17702088
|
Notrax
411S0323BL |
91 cm | 60 cm |
28.75 €
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
17742088
|
Notrax
411S0335BL |
150 cm | 91 cm |
66.05 €
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
17782068
|
Notrax
411C0324BL |
1830 cm | 60 cm | ||||||
17722078
|
Notrax
411C0336BL |
1830 cm | 91 cm | ||||||
17762078
|
Notrax
411C0348BL |
1830 cm | 122 cm | ||||||
Description
- Pebble pattern provides non-directional traction and is easy to clean
- Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
- All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Black | |
Microcellular Vinyl | |
Industrial and Commercial Matting |