missing translation for 'onlineSavingsMsg'
Learn More

American Educational Products Solar Furnace

A working model of an industrial solar furnace.

Brand:  American Educational Products 4701

Product Code. 13176350

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes:

Teacher's guide

This item is not returnable. View return policy

Description

Description

  • Furnace uses a 12 in. diameter (30.5cm), 7.3 in. (18.5cm) focal length parabolic dish
  • Concentrates solar radiation to a copper container that collects and concentrates the radiant heat
  • Unit disassembles easily for storage

Order Info

Dimensions: 12 x 15 in. (30.5 x 38.1cm).

Specifications

Specifications

3 to 12
12 in.
7.3 in.
Teachers guide
30.5 cm
18.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
American Educational Products Solar Furnace > Solar Furnace

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.