missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Solaris 2000 R Small Incubated and Refrigerated Benchtop Orbital Shaker
GREENER_CHOICE

Thermo Scientific™ Solaris Incubated and Refrigerated Benchtop Shakers have an intuitive touch screen display, programmability, and multi-language support. Peltier technology provides refrigerant-free cooling with quiet operation.

Brand:  Thermo Scientific™ SK2002

 View more versions of this product

Product Code. 17899484

  • 9768.60 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket

Description
Show More
Specifications

Specifications

25 mm
15 to 525 rpm, ±1 rpm
100–240 V, 50/60 Hz
Orbital
35 lb.
12 x 14 in.
Solaris SK2000 R
11 x 14 in. platform
CFC free
16 kg
100–240 V, 50/60 Hz
5°C to 60°C
Shaker
CE Cert, UL (Canada/US) File E311968, UK Mark
27.6 x 14.2 x 18.1 in.
Touchscreen
Standard Warranty includes 2 years labor, 5 years parts, and 10 years on the drive mechanism
Less hazardous, energy efficient
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Solaris 2000 R Small Incubated and Refrigerated Benchtop Orbital Shaker >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.