missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Solvex™ 37-900 Series Red Nitrile Immersion Gloves

Offers greater strength and superior chemical resistance. Ansell SolvexTM 37-900 Series Red Nitrile Immersion Gloves set new standards for mechanical performance, with unrivalled abrasion and puncture resistance - almost twice that of the current leading nitrile.

Brand:  Ansell™ 37900110BP

87.77 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.


 View more versions of this product

Product Code. 12425170

  • 110.00 € / 12 pairs
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Out-performs all existing nitriles in permeation tests
  • Every glove individually air-pressure tested
  • Every glove subjected to a leakproof test
  • ISO 9001 certified
  • Superior standards of safety for high-risk chemical handling
  • Ideal for oil and petrol refining, laboratory analysis, cleaning and the printing industry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nitrile
Level - 0
Immersion Gloves
37-900
Red
Flocked
Abrasion, puncture and superior chemical resistance
Nitrile
Yes
11
0.42 mm
Straight
1 Pair
37-900
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.