Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Rectangular Buckets
For use with four-place swinging bucket rotor 75-002-000 when equipped with rectangular bucket 75-002-001
Brand:  Thermo Scientific™ 75002021
 View more versions of this product
Product Code. 10216842
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For use with four-place swinging bucket rotor 75-002-000 when equipped with rectangular bucket 75-002-001
Adapters sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15 mL Blood Collection Tube
4-place Swing rotor with rectangle bucket