Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Rectangular Buckets
For use with four-place swinging bucket rotor 75-002-000 when equipped with rectangular bucket 75-002-001
107.54 € - 193.00 €
Product Code | Brand | Tube Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Type | Price | Quantity & Availability | |||||
10140941
|
Thermo Scientific™
75005322 |
15 mL DIN |
107.54 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10216842
|
Thermo Scientific™
75002021 |
15 mL Blood Collection Tube |
193.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10059430
|
Thermo Scientific™
75005386 |
Conical |
172.86 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For use with four-place swinging bucket rotor 75-002-000 when equipped with rectangular bucket 75-002-001
Adapters sold individually; order correct quantity to equip rotor.
Specifications
4-place Swing rotor with rectangle bucket |