Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Buckets for 4-Place Swinging Bucket Rotors
Round or rectangular buckets for use with Four-Place Swinging Bucket Rotors 75006445 and 75006434
Brand:  Thermo Scientific™ 75006441
 View more versions of this product
Product Code. 10458191
  • 472.35 € / Each
Explore more special offers
Description
Description
Order Info
Sold individually; order correct quantity to equip rotor. Sealing lids sold separately. Accessory adapters accept assorted tube styles and volumes from 1.5 to 250mL.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Swinging bucket rotor
Round Bucket; Capacity: 1 x 750 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products