missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Space-Saver Desiccators
For dry storage and protection of moisture-sensitive products.
62.55 € - 137.00 €
Product Code | Brand | Volume (English) | Max. Clearance Above Plate | Diameter (Metric) Inner | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (English) | Max. Clearance Above Plate | Diameter (Metric) Inner | Price | Quantity & Availability | |||
11872722
|
Bel-Art™
F42031-0000 |
0.20 cu. ft. | 15.7 cm | 19.7 cm |
97.50 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||
11862722
|
Bel-Art™
F42030-0000 |
0.09 cu. ft. | 12.1 cm | 14.9 cm |
62.55 €
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||
11882722
|
Bel-Art™
F42032-0000 |
0.31 cu. ft. | 19.7 cm | 24 cm |
137.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||
Description
- Strong and shatterproof with extra heavy walls
- Flat dome, clear polycarbonate top maximizes interior clearances and provides greater interior volume
- Large ribbed knob for easy lifting of top
- Includes 0.32cm (0.125 in.) thick perforated plate
- Disposable or reusable desiccant can be placed on or under perforated plate
- Neoprene O-ring ensures tight seal to 0°C without need for grease
- White polypropylene bottom supports incandescent crucibles on High Heat Minerit HD Ceramic Desiccator Plates
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title