missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Capacity (Metric) | Shelves | Temperature Range | Style | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Shelves | Temperature Range | Style | Price | Quantity & Availability | ||||||
11528954
|
Labcold™
RLCF0720 |
215 L | 1 Basket | -18°C to -25°C | Chest |
4170.00 €
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
11538954
|
Labcold™
RLCF1520 |
447 L | 2 Baskets | -18°C to -25°C | Chest |
4330.00 €
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
11548954
|
Labcold™
RLCF2120 |
607 L | 1 Basket | -18°C to -25°C | Chest |
5645.00 €
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
15523851
|
Labcold™
RLVF1514 |
400 L | 6 Adjustable | -12°C to -24°C | N/A | N/A | N/A | ||||||
Description
- CFC and HCFC free
- Operator controlled defrost
- Alarm/temperature display can be fitted as an optional extra
- Chest freezers feature integral lid lock
- Locks available for all upright freezers as an additional extra
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Labcold™ Sparkfree Freezers