missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices

Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)

Brand:  Spectrum™ G235060

193.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11571170

  • 284.00 € / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)

Available in volume sizes: 1mL, 5mL and 10 mL. Resealable screw cap is color-coded for 9 available MWCOs and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5mL and 10mL sizes). Flotation ring (included) provides buoyancy to maximize dialysis efficiency.

  • Superior handling and leak protection
  • Resealable screw cap and pipet for easy loading and total recovery
  • Highest membrane purity, no heavy metals and sulfides
  • Minimizes volume specific dilution
  • 9 MWCOs ranging from 100 to 1,000,000 Daltons

Specifications

Specifications

Float-A-Lyzer G2 Dialysis Device
Cellulose Ester
0.9 in.
23 mm
100 mm
Screw Cap
Disposable pipette for 5 and 10mL, flotation ring
Ready to use laboratory dialysis devices, highest membrane purity, superior handling and leak prevention, 95 to 98% sample recovery, perfect for dialyzing electro-eluted proteins or chromatography fractions
300 kDa
0.39 in.
10 mm
3.9 in.
Polypropylene
Amber
12 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices > 300 kd MWCO; Amber; 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.