missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ RC Dialysis Discs

Brand:  Spectrum™ 132480

256.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10577124

  • 263.00 € / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Material: Regenerated Cellulose
  • Hydrophilic
  • Symmetric porosity
  • Carries no fixed charge
  • Does not adsorb most solutes
  • pH limits: 2 to 12
  • Packaged dry with glycerol (must be rinsed out with water)

For laboratory dialysis functions such as desalting, buffer exchange or molecular separation

Specifications

Specifications

RC Dialysis Disc
Spectra/Por 2
47 mm
50 Pack
12 to 14 kDa
Regenerated Cellulose
1.85 in.
For laboratory dialysis functions such as desalting, buffer exchange or molecular separation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Spectrum™ Spectra/Por™ RC Dialysis Discs > Spectra/Por 2; MWCO: 12 to 14 kD; Diameter: 47mm (1.85 in.)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.