missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra/Por™ RC Dialysis Discs
263.00 € - 328.00 €
Specifications
Product Type | RC Dialysis Disc |
---|---|
Material | Regenerated Cellulose |
Quantity | 50 Pack |
For Use With (Application) | For laboratory dialysis functions such as desalting, buffer exchange or molecular separation |
Product Code | Brand | MWCO | Diameter (Metric) | Membrane | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Diameter (Metric) | Membrane | Price | Quantity & Availability | |||
12628273
|
Spectrum™
132478 |
6 to 8 kDa | 33 mm | Spectra/Por 1 |
328.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10577124
|
Spectrum™
132480 |
12 to 14 kDa | 47 mm | Spectra/Por 2 |
263.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Material: Regenerated Cellulose
- Hydrophilic
- Symmetric porosity
- Carries no fixed charge
- Does not adsorb most solutes
- pH limits: 2 to 12
- Packaged dry with glycerol (must be rinsed out with water)
For laboratory dialysis functions such as desalting, buffer exchange or molecular separation
Specifications
RC Dialysis Disc | |
Regenerated Cellulose | |
50 Pack | |
For laboratory dialysis functions such as desalting, buffer exchange or molecular separation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spectrum™ Spectra/Por™ RC Dialysis Discs