Learn More
American Educational Products Spectroscope Kit
An excellent analysis of sunlight
Brand:  American Educational Products VTN12302478
Product Code. 13122990
  • 2.93 € / Each
Estimated Shipment: 07-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This diffraction grating spectroscope is used to show that sunlight can be broken into colored light, and to study these colors and their relationships.
•    Students view different types of light and compare their spectra
•    By drawing the colors side-by-side and to the same scale, students can form their own spectrum
•    Observe and define different spectra
•    An excellent preparation for understanding methods in astronomy
Specifications
Specifications
Escp Spectroscope Kit
6 to 12
11 to 18
Used to show that sunlight can be broken into colored light, and to study these colors and their relationships
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
American Educational Products Spectroscope Kit > Escp Spectroscope Kit