Learn More
Spermine, 97%, Thermo Scientific Chemicals
55.98 € - 154.77 €
Chemical Identifiers
CAS | 71-44-3 |
---|---|
Molecular Formula | C10H26N4 |
Molecular Weight (g/mol) | 202.35 |
MDL Number | MFCD00008215 |
InChI Key | PFNFFQXMRSDOHW-UHFFFAOYSA-N |
Synonym | spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl |
PubChem CID | 1103 |
ChEBI | CHEBI:15746 |
IUPAC Name | N,N'-bis(3-aminopropyl)butane-1,4-diamine |
SMILES | NCCCNCCCCNCCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10795781
|
Thermo Scientific Acros
132750010 |
1 g | Glass bottle |
55.98 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10488480
|
Thermo Scientific Acros
132750050 |
5 g | Glass bottle |
154.77 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71-44-3 | |
202.35 | |
PFNFFQXMRSDOHW-UHFFFAOYSA-N | |
1103 | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine |
C10H26N4 | |
MFCD00008215 | |
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl | |
CHEBI:15746 | |
NCCCNCCCCNCCCN |
Specifications
71-44-3 | |
0.9370g/mL | |
110°C | |
C10H26N4 | |
MFCD00008215 | |
15, 8870 | |
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl | |
NCCCNCCCCNCCCN | |
202.35 | |
CHEBI:15746 | |
97% |
26.0°C to 30.0°C | |
130.0°C (0.1 mmHg) | |
97% | |
H2N(CH2)3NH(CH2)4NH(CH2)3NH2 | |
04, II, 704 | |
0.937 | |
PFNFFQXMRSDOHW-UHFFFAOYSA-N | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine | |
1103 | |
202.34 | |
Spermine, 97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-754-2
missing translation for 'rtecsNumber' : EJ7175000
missing translation for 'tsca' : TSCA
- Spermine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only