missing translation for 'onlineSavingsMsg'
Learn More

SPEX CertiPrep™ Accessories for Pipet Washer and Dryer

Accessories for use with Pipet Washer and Dryer

190.00 € - 291.00 €

Specifications

For Use With Pipet Washers
For Use With (Equipment) For pipet washer and dryer
View More Specs

Products 3
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
11466068
View Documents
SPEX CertiPrep™
PIPBASIN-1
Pipet Washer Basin
190.00 €
1 unit
Estimated Shipment: 24-06-2025
to see stock.
Add to basket
 
15236116
View Documents
SPEX CertiPrep™
PIPPUMP-115V
Pipette Washer Pump, 115V
243.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
13206359
View Documents
SPEX CertiPrep™
PIPPUMP-230V
Pipette Washer Pump, 230V
291.00 €
1 unit
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
 
Description

Description

Pump and Basin both constructed of durable polyethylene.

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Specifications

Specifications

Pipet Washers
For pipet washer and dryer
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SPEX CertiPrep™ Accessories for Pipet Washer and Dryer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.