missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Spill Tray

Spill Tray allows you to store your small containers in a confined space.

Brand:  Gilac™ G154421-3

 View more versions of this product

Product Code. 17968721

  • 12.55 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Allows you to store your small containers in a confined space.
  • Rounded corners for easier cleaning.
  • Machine washable.
  • Other colors available on request.
  • To choose the right size of spill tray, follow the legal requirements*in force: The storage of any liquid likely to create water or soil pollution is associated with a retention capacity whose volume is equal to the greater of the following two values:
    • 100% of the capacity of the largest container,
    • 50% of the capacity of the associated containers.

* The order of 19 July 2011 (Article 3) amending the order of 4 February 1998 (Article 10) on the prevention of accidental risks in installations classified for environmental protection subject to authorisation.

Specifications

Specifications

8 L
75 mm
HDPE
335 mm
3 in.
2.1 gal.
White
485 mm
Utility Tray
19.1 in.
13.2 in.
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers