Learn More
Cytiva Spin Adapters for Gravity Columns
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
Brand:  Cytiva 28-9232-43
 View more versions of this product
Product Code. 11797819
  • 15.00 € / Pack of 10
Estimated Shipment: 06-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Convenient adapters, enable spinning of all gravity columns.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
10 Pack
Spin Column Adapter
Polypropylene
Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD MiniTrap G-25 column for centrifugation