Learn More
Cytiva Spin Adapters for Gravity Columns
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
15.00 € - 19.00 €
Specifications
Description | Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns. |
---|---|
Material | Polypropylene |
Quantity | 10 Pack |
Type | Spin Column Adapter |
Product Code | Brand | Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Application) | Price | Quantity & Availability | ||||
11717829
|
Cytiva
28-9232-45 |
Spin Column Adapter | Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD-10 desalting column for centrifugation |
19.00 €
Pack of 10 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
11707829
|
Cytiva
28-9232-44 |
Spin Column Adapter | Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD MidiTrap G-25 column for centrifugation |
19.00 €
Pack of 10 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
11797819
|
Cytiva
28-9232-43 |
Spin Column Adapter | Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD MiniTrap G-25 column for centrifugation |
15.00 €
Pack of 10 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
Description
- Convenient adapters, enable spinning of all gravity columns.
Specifications
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns. | |
Polypropylene | |
10 Pack | |
Spin Column Adapter |