Learn More
Lenz Laborglasintrumente Spiral Condensers, with Glass Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05233504
 View more versions of this product
Product Code. 10737331
  • 179.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Spiral condensers for cooling condensates
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Specifications
500 mm
NS 29/32