missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Spiral Condensers, with Glass Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
168.00 € - 179.00 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10601902
|
Lenz Laborglasintrumente
05233404 |
400 mm |
168.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
10737331
|
Lenz Laborglasintrumente
05233504 |
500 mm |
179.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
Description
- Spiral condensers for cooling condensates
- With NS socket and NS cone
- Water connection: glass olives
- Made of DURAN™ tubing
Specifications
NS 29/32 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title