missing translation for 'onlineSavingsMsg'
Learn More

Cole Parmer™ Square Polystyrene Weigh Boats, White, 100/Pk

Rounded corners simplify transfer of powdered or granular samples.

Brand:  Cole Parmer™ 01017-04

 View more versions of this product

Product Code. 15406033

  • 16.35 € / Pack of 100
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Rounded corners facilitate sample transfer.
  • Disposable.
  • White translucent color dishes for traditional applications.
  • Translucent, rubberized polystyrene withstands temperatures to 180°F, flexes for easy pouring.
  • Biologically inert; resistant to dilute acids, aqueous solutions, alcohols, bases.
  • For use with balances, microwave moisture analyzers, hundreds of other sample handling applications.
  • Flat bottom resists tipping.
  • Contoured sides permit no-slip handling.
Specifications

Specifications

Flat
White
Polystyrene
Weighing Boat
180°F
4.1 cm
0.8 cm
20 mL
Single-use
Square
100/Pk.
1.625 in.
0.313 in.
0.313 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers