Learn More
Corning™ PYREX™ Squibb Glass Separatory Funnel with PTFE Valve and Threaded Screw Cap
Constructed of durable PYREX borosilicate glass
Brand:  Corning™ 10-462-583
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12700858
  • 174.90 € / Each
Estimated Shipment: 04-09-2024
to see stock.
Add to basket
Description
Description
Improved Squibb type separatory funnel provides an unbreakable, all PTFE, valve and drip tip and has a screw thread closure at the top. Supplied complete with PTFE valve assembly and screw cap with PTFE faced silicone liner.
For glass stopper only, see Cat. No. 7650. For poly stopper only, see Cat. No. 7624.
Specifications
Specifications
250 mL
Borosilicate Glass
1/Cs.
Squibb Separatory Funnel