Learn More
Corning™ PYREX™ Squibb Glass Separatory Funnel with PTFE Valve and Threaded Screw Cap
Constructed of durable PYREX borosilicate glass
174.90 €
Specifications
Quantity 1/Cs.
View More Specs
Products 1
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
12700858
View Documents Promotion Details
Corning™
10-462-583
250 mL
174.90 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
Description
Description
Improved Squibb type separatory funnel provides an unbreakable, all PTFE, valve and drip tip and has a screw thread closure at the top. Supplied complete with PTFE valve assembly and screw cap with PTFE faced silicone liner.
For glass stopper only, see Cat. No. 7650. For poly stopper only, see Cat. No. 7624.
Specifications
Specifications
1/Cs.