Learn More
Sampling Systems™ Stainless Powder Funnel
Sampling Systems™ Stainless Powder Funnel are made from high quality Stainless Steel and suitable for hygienic and pharmaceutical applications.
Brand: Sampling Systems™ A716-200
Description
- High quality 316L Stainless Steel
- Satin finish
- Range of sizes available
- Tri-ferrule fitting at spout base (BS4825-3)
Specifications
200mm | |
Stainless Steel | |
1.5 in. ferrule on spout |
38mm | |
230 mm | |
Funnel |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.