Learn More
Scotsman™ Stainless Steel AF103 Ice Flaker
Brand:  Scotsman™ AF103
Product Code. 12682645
  • 5955.00 € / Each
Explore more special offers
Description
Description
Ice flakes are extruded at a temperature just below zero degrees and contain 20% residual water content. The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access. Electrical supply 230V/50Hz.
  • Up to 108kg (238 lbs.) of flake ice production per day
  • Up to 30kg (66 lbs.) built-in ice storage
  • Electronically controlled operation, with self-diagnostic functions with external alarm lights providing operational information for early alert and fast diagnosis of operating issues
  • External components constructed from corrosion-resistant stainless steel crafted for optimal aesthetic appeal through superior fit and finish
  • Ergonomically convenient, slanted & disappearing storage bin door
  • Front access for easier maintenance
  • Front-in, front-out airflow allows for built-in installations
  • CFC-free R-404A refrigerant
  • 120mm (4 12/16 in.) legs included, extensions available as option
  • Two years manufacturer warranty Scotsman flaked ice is ideal for a variety of applications within laboratory and industrial environments.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Scotsman™ Stainless Steel AF103 Ice Flaker > Capacity: 30kg