Learn More
Hamilton™ Stainless Steel Anion Exhchange Columns
Brand:  Hamilton™ 79734
 View more versions of this product
Product Code. 10452543
  • 1105.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.11 μeq/g
0.176 meq/column
PRP-X110
PSDVB/Trimethylammonium
5°C to 60°C
250 mm
4.1 mm
PRP-X110
7 μm
Stainless Steel
100 &‌Aring;
L47
4.1 mm