Learn More
Hamilton™ Stainless Steel Anion Exhchange Columns
Brand:  Hamilton™ 79365
 View more versions of this product
Product Code. 11503504
  • 764.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.19 μeq/g
10 μm
Stainless Steel
100 &‌Aring;
4.1 mm
PRP-X100
PRP-X100
PSDVB/Trimethylammonium
50 mm
4.1 mm