Learn More
IKA™ Stainless Steel Beater
Brand:  IKA™ 0002983000
288.58 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 10449602
  • 276.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Beater
For Pulverizing Substances with a Mohs Hardness up to 9
Up to 9 °Mols
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IKA™ Stainless Steel Beater > For Use With: Pulverizing Substances with a °Mohs Hardness up to 9