Learn More
RETSCH Stainless Steel Bottom Sieve For SK100 Cross Beater Mills
Brand: RETSCH 02.407.0001
This item is not returnable.
View return policy
Specifications
1Each | |
Bottom Sieve | |
SK100 Cross Beater Mills |
250 μm | |
Stainless Steel |
Product Content Correction
Product Title
RETSCH Stainless Steel Bottom Sieve For SK100 Cross Beater Mills > Pore Size: 250 μm
Spot an opportunity for improvement?Share a Content Correction