Learn More
Sampling Systems™ Stainless Steel Cable
Sampling Systems™ Stainless Steel Cable is designed to be used in conjuction with a Disposable Bailer with or without a weighted end.
Brand: Sampling Systems™ 5070D-10
This item is not returnable.
View return policy
Description
- Cable for Disposable Bailer
- PTFE coated Stainless Steel
- Avalable in a variety of lengths
Specifications
Cable | |
PTFE | |
10 m |
Stainless Steel | |
Disposable Bailer |