Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class M1 with certificate
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
Brand:  Fisherbrand™ FB67846
 View more versions of this product
Product Code. 15555129
  • 501.00 € / 1 unit
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
M1
1 g to 1 kg
1 g to 1 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class M1 with certificate > 1g to 1kg