Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class M1
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
Brand: Fisherbrand™ FB67835
Description
- Class M1 stainless steel calibration weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
M1 | |
1 to 100 g | |
1 g to 100 g | |
Cylindrical |
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class M1 > 1g to 100g
Spot an opportunity for improvement?Share a Content Correction