Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class M1
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
134.00 € - 395.00 €
Specifications
Classification M1
Shape Cylindrical
View More Specs
Products 2
Product Code Brand Weight (Metric) Price Quantity & Availability  
Product Code Brand Weight (Metric) Price Quantity & Availability  
15545119
View Documents
Fisherbrand™
FB67835
1 to 100 g
134.00 €
1 unit
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
15545129
View Documents
Fisherbrand™
FB67845
1 g to 1 kg
395.00 €
1 unit
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Description
Description
  • Class M1 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
M1
Cylindrical
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class M1