missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Cylindrical Scoops

Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Cylindrical Scoops are an ideal accessory to have if you own a weighing scale.

Brand:  Fisherbrand™ 9030

 View more versions of this product

Product Code. 11573452

  • 31.95 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Cylindrical Scoops are an ideal accessory to have if you own a weighing scale.

  • Durable, stainless steel, highly polished, and easy to clean cylindrical scoop 
  • Can be sterilised by autoclaving
  • Well-suited for pharmaceutical and environmental sampling
  • Ideal for weighing and handling of samples and other materials
  • Great accessory to have if you own a weighing scale
  • Perfect for weighing anything from liquid to gems and other materials
Specifications

Specifications

Scoop
Autoclavable
Sterile
325 mm
Stainless Steel
500 mL
Cylindrical
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.