Learn More
asecos™ Stainless Steel Dispensing Canister
The Stainless Steel Dispensing Canister is ideal for the safe distribution and dosing of flammable liquids.
227.00 € - 466.00 €
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
17866386
|
asecos™
14757 |
5 L | 126.00 mm |
344.00 €
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
17876496
|
asecos™
14745 |
1 L | 210.00 mm |
227.00 €
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
17806546
|
asecos™
14746 |
2.5 L | 280.00 mm |
236.00 €
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
17846336
|
asecos™
14747 |
5 L | 445.00 mm |
268.00 €
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
17846546
|
asecos™
14758 |
10 L | 240.00 mm |
376.00 €
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
17816456
|
asecos™
14759 |
20 L | 495.00 mm |
466.00 €
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
Description
- With fine dosing tap made of stainless steel, incl. ventilation, PTFE seals, integrated flame arrester (removable) and plastic protective ring
- Stainless Steel
Specifications
Gray | |
Stainless Steel |