missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Laboratory Tray

Manufactured by Bochem™, the Stainless Steel Laboratory Tray is useful for containing and carrying items used for experimentation in the lab.

Brand:  Bochem™ 8703

 View more versions of this product

Product Code. 10170261

  • 127.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless Steel
  • Round Angles
  • Steel sides to protect contents
Specifications

Specifications

355 x 305 mm
60 mm
Stainless Steel
260 mm (Bottom), 305 mm (Top)
Rectangular
355 x 305 x 60 mm
320 mm (Bottom), 355 mm (Top)
Laboratory Tray
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.