missing translation for 'onlineSavingsMsg'
Learn More
Learn More
De Buyer™ Stainless Steel Ladle with Ergonomic Handle
Grip the perfectly angled ergonomic handle shape on the De Buyer™ Stainless Steel Ladle with Ergonomic Handle that lets you insert your thumb.
16.00 € - 270.00 €
Product Code | Brand | Volume (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
11967435
|
De Buyer™
3021.20 |
2.06 L | 635 mm |
270.00 €
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
11973674
|
De Buyer™
3021.08 |
0.13 L | 340 mm |
16.00 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
11903684
|
De Buyer™
3021.14 |
0.67 L | 140 mm |
47.00 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11947435
|
De Buyer™
3021.16 |
1 L | 430 mm |
170.00 €
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
Description
- Wide and rigid 2.5mm handle
- Fixed hook at end of handle
- Litres capacity engraved on handle
- Dishwasher safe
Specifications
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
De Buyer™ Stainless Steel Ladle with Ergonomic Handle