missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Lids For Test Sieve

Providing a secure closure to Fisherbrand™ Stainless Steel Test Sieves.

Brand:  Fisherbrand™ 01/100SBW LID

 View more versions of this product

Product Code. 11592573

  • 41.05 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Available in 100 and 200mm diameter sizes
  • Conforms to BS 410: Part 1: 2000, ISO 3310: Part 1: 2000
Specifications

Specifications

1 Ea.
Stainless Steel
100 mm
Sieve Cover
Providing closure to test sieves
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers