missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
11346680
|
Sartorius™
YCW5128-02 |
100 g |
248.00 €
Each |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||||
11316680
|
Sartorius™
YCW4228-02 |
20 g |
204.00 €
Each |
Estimated Shipment: 18-06-2025
Log in to see stock. |
||||||
11336680
|
Sartorius™
YCW4528-02 |
50 g |
221.00 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
11461947
|
Sartorius™
YCW5228-02 |
200 g |
285.00 €
Each |
Estimated Shipment: 18-06-2025
Log in to see stock. |
||||||
Description
- Made of special stainless-steel
- Density: 7,950kg/m3
- Tolerances comply with OIML R111-1
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sartorius™ OIML Class E2 Cylindrical Weights with DAkkS calibration certificate