missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Stainless Steel OIML Class F2 Calibration Weight Without DAkkS Calibration Certificate

Highly polished steel calibration weights from 100g to 10kg

Brand:  Sartorius™ YCW6548-00

638.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10012121

  • 656.00 € / Each
Estimated Shipment: 18-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

To OIML Class E2, F1 and F2 specification which includes DKD (Deutscher Kalibrierdienst accredited by the PTB) certificate (UKAS Equivalent).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Plastic
5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius™ Stainless Steel OIML Class F2 Calibration Weight Without DAkkS Calibration Certificate > Mass: 5kg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.