Learn More
De Buyer™ Stainless Steel Pastry Bowls
Brand:  De Buyer™ 3250.24
 View more versions of this product
Product Code. 11910605
  • 57.00 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Pastry Bowl
For making mixtures, mixing, Whisking, heating by bain-marie, for storing a preparation or ingredients
112 mm
Flat Bottom
3.5 L
Stainless Steel
Fridge, deep-freezer, blast-chiller and oven
Each
Sterile