Learn More
Nickel Electro™ Stainless Steel Perforated Tray
Nickel Electro™ Stainless Steel Perforated Tray is to be used with flask clips and can be used in a variety of baths.
Brand:  Nickel Electro™ SFT9000
 View more versions of this product
Product Code. 11419549
  • 407.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Supplied ready to assemble
  • Stainless steel construction
  • Suits glass/polypropylene Erlenmeyer flasks
  • Tray simply drops onto shaker or rocking table platform and locates snugly
Specifications
Specifications
Universal Spring Tray
Universal spring tray
2 x 1000 mL, 6 x 500 mL, 15 x 100 mL, 35 x 25 mL
Stainless Steel
NE5-28 Shaker Baths