Learn More
Nickel Electro™ Stainless Steel Perforated Tray
Nickel Electro™ Stainless Steel Perforated Tray is to be used with flask clips and can be used in a variety of baths.
Brand:  Nickel Electro™ SFT0500
 View more versions of this product
Product Code. 12728978
  • 341.00 € / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Supplied ready to assemble
  • Stainless steel construction
  • Suits glass/polypropylene Erlenmeyer flasks
  • Tray simply drops onto shaker or rocking table platform and locates snugly
Specifications
Specifications
Perforated Tray
Yes
8
Stainless Steel
NE5-28 Shaker Baths