missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments Stainless Steel Radial Flow Impeller

Precise stirring requires careful overhead stirrer selection. This Stainless Steel Radial Flow Impeller is ideal for little or average viscosity stirring at average speeds.

Brand:  Heidolph Instruments 509-20000-00

Product Code. 10569521

  • 108.00 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Average speed stirring
  • Mixing tasks with little or average viscosity
  • Ideal for gassing of liquids
  • Creates a radial flow
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.3 in.
8 mm
400 mm
Radial Flow Impeller
8 mm
15.7 in.
2000 rpm
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heidolph Instruments Stainless Steel Radial Flow Impeller > Radial Flow Impeller; Length: 400 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.